Learn More
Sciencell Research Laboratories STECM PHENOL RED FREE 500ML
Catalog No. NC1588992
Encompass
Stellate Cell Medium. Phenol red free (no supplements) 500ml; SteCM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of stellate cell growth supplement (SteCGS, Cat. No. 5352) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).
Catalog No. NC1588992 Supplier Sciencell Research Laboratories Supplier No. 5301BPRF
Stellate Cell Medium. Phenol red free (no supplements) 500ml; SteCM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of stellate cell growth supplement (SteCGS, Cat. No. 5352) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).