Learn More
Grainger Steel 11 D 14.1 H 11.1 W Inline Duct Fan
Catalog No. 502750150
Encompass_Preferred
The Tjernlund In-Line Duct Fan 160 cfm Max is a duct fan designed for inline installation to provide airflow in ventilation systems. It features a 1/3 HP PSC motor with permanently lubricated ball bearings for continuous duty, thermally protected with automatic reset. The fan has a 6" diameter and is constructed of galvanized steel housing. It can deliver up to 160 cfm at 0.0" static pressure. The maximum static pressure is 0.50" H2O. This duct fan has a 4-speed rotary switch and is ETL listed for US and Canada. The overall dimensions are 11-1/4" length x 9-3/8" width x 12-3/4" height. It weighs approximately 13 lbs.
Catalog No. 50-275-0150 Supplier Grainger Supplier No. 14U160
The Tjernlund In-Line Duct Fan 160 cfm Max is a duct fan designed for inline installation to provide airflow in ventilation systems. It features a 1/3 HP PSC motor with permanently lubricated ball bearings for continuous duty, thermally protected with automatic reset. The fan has a 6" diameter and is constructed of galvanized steel housing. It can deliver up to 160 cfm at 0.0" static pressure. The maximum static pressure is 0.50" H2O. This duct fan has a 4-speed rotary switch and is ETL listed for US and Canada. The overall dimensions are 11-1/4" length x 9-3/8" width x 12-3/4" height. It weighs approximately 13 lbs.