Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Steel
SCI_ED

Catalog No. S25587 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Steel Shot
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25587 500 g Steel Shot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25587
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Specifications

Color Gray
Chemical Name or Material Steel
Quantity 500 g
Packaging Steel Shot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.