Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Dialysis Tubing
SCI_ED

Catalog No. S02460 Shop All Aldon Products
Click to view available options
Diameter (Metric):
75 mm
Length (English):
100 ft.
Width (Metric):
47.7 mm

Catalog No. S02460

Supplier: Innovating Science™ IS35049

Only null left
Add to Cart
Add to Cart

Dialysis tubing of various sizes.

The semi-permeable membrane allows for selective transport within a solution.

Specifications

Product Type Dialysis Tubing
MWCO 7 kDa
Format Tubing
Diameter (Metric) 75 mm
Diameter (English) 2.95 in.
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (Metric) 47.7 mm
Sample Volume (Metric) 2.2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.