Learn More
World Precision Instrument Steel Base Plate, 32 lbs
SDP
Catalog No. 50822960
Encompass
Base plate - Mounting surface for magnetic stands
A magnetic stand requires a steel mounting serface. WPI's steel base plates have plenty of mass to give stability to your experimntal setup. Beveled edges make them easy to handle. Rubber feet hold them off the benchtop, making them easier to grasp when you are moving them. The special black coating provides a durable, protective, non-reflective finish.
Catalog No. 50-822-960 Supplier World Precision Instrument Supplier No. 5479
Base plate - Mounting surface for magnetic stands
A magnetic stand requires a steel mounting serface. WPI's steel base plates have plenty of mass to give stability to your experimntal setup. Beveled edges make them easy to handle. Rubber feet hold them off the benchtop, making them easier to grasp when you are moving them. The special black coating provides a durable, protective, non-reflective finish.