Learn More
Research Products International Corp Disruption Beads for Fibrous Plant Tissue, 3.2mm Bead Size, Chrome Steel Beads, 7.9 g/cc Density, 1 Pound Bottle
SDP
Catalog No. 50998373
Encompass
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.
Catalog No. 50-998-373 Supplier Research Products International Corp Supplier No. 9840
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.