Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Steel Beakers With Rim, Low Form

Catalog No. 21124617 Shop All Surgical Design Products
Change view
Click to view available options
Capacity (Metric):
32 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-124-617 32 mL
21-124-610 100 mL
21-124-611 250 mL
21-124-612 500 mL
21-124-613 1 L
21-124-614 2 L
21-124-615 3 L
21-124-616 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 21-124-617 Supplier Surgical Design™ Supplier No. 785142
Add to Cart
Add to Cart

Low form steel beakers with rim deliver durability and chemical resistance for heating, mixing, or reagent handling.

  • Manufactured from premium corrosion-resistant stainless steel for maximum durability and longevity
  • Low form design provides stability and minimizes spillage during heating or mixing
  • Rolled rim edge ensures easy handling and smooth pouring of liquids
  • Uniform wall thickness supports even heat distribution and resistance to denting
  • Non-reactive surface safe for use with most reagents and solutions
  • Autoclavable and reusable, ideal for sterilized laboratory environments
  • Available in capacities from 32 ml to 5000 ml to meet diverse lab needs
  • Lightweight yet sturdy construction for dependable daily use
  • Suitable for general lab, educational, chemical, and industrial environments
  • Ideal for sample preparation, solution heating, reagent mixing, and material storage

Specifications

Capacity (Metric) 32 mL
Material Stainless Steel
Type Steel Beakers
Graduated No
Style Griffin, Low Form
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.