Learn More
Fine Science Tools Burrs for Micro Drill, 2.1mm, 10/pk
SDP
Catalog No. NC9685727
Encompass
Tip Diameter: 2.1mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No
Catalog No. NC9685727 Supplier Fine Science Tools Supplier No. 1900721
Tip Diameter: 2.1mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No