Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Steel Containment Pallet

Catalog No. 19103122 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
1 drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-103-122 1 drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-122 Supplier Brady™ Supplier No. SS1DR

Comply with governmental secondary containment regulations or get the security of knowing your facility is protected. Brady™ spill pallets offer an extra level of protection.

  • Whether you are looking to comply with governmental secondary containment regulations or just want the extra security of knowing your facility is protected, spill pallets offer an extra level of protection
  • Offers ample capacity for containing potentially hazardous leaks
  • Galvanized steel build is corrosion resistant and forklift compatible

Specifications

Color Silver
Material Galvanized Steel
For Use With (Equipment) 1 drum
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.