Learn More
Thwing Albert Instrument Co STEEL DIE WEAR PLATE 12" X 12", RECOMMENDED FOR THE 240-7B CUTTER ONLY
SDP
Catalog No. NC3135933
Encompass
Recommended for thin materials, such as plastic film, tissue, thin papers. The plate will be installed ifordered with a new 240-7B Cutter.
Catalog No. NC3135933 Supplier Thwing Albert Instrument Co Supplier No. 002410100
Recommended for thin materials, such as plastic film, tissue, thin papers. The plate will be installed ifordered with a new 240-7B Cutter.