Learn More
Electron Microscopy Sciences Brush with Steel Ferrules
SDP
Catalog No. 5030358
Encompass_Preferred
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax. They are offered with metal ferrules for improved durability. The overall length is 4 1/4" long, and the brush is 1/8" wide.
Material: Steel Ferrules
Catalog No. 50-303-58 Supplier Electron Microscopy Sciences Supplier No. 6558201
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax. They are offered with metal ferrules for improved durability. The overall length is 4 1/4" long, and the brush is 1/8" wide.
Material: Steel Ferrules