Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Steel Grip Jacket, Non-Hazardous Protection, X-Large, Cotton, 1 ea

Catalog No. 5000673970
Encompass

This green cotton jacket provides non-hazardous protection. It features a snaps closure and is designed for a chest size of 48 to 50 in., including one pocket.

  • Garment style: jacket
  • Size: x-large
  • Garment type: flame-resistant, welding, flame-retardant
  • Color: green
  • Material: cotton
  • Closure type: snaps
  • Hazardous protection level: non-hazardous protection
  • Chest size: 48 to 50 in.
  • Material weight: 9.00 oz
  • Number of pockets: 1
  • Country of origin: Mexico

Catalog No. 50-006-73970 Supplier MSC Supplier No. 66550559
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This green cotton jacket provides non-hazardous protection. It features a snaps closure and is designed for a chest size of 48 to 50 in., including one pocket.

  • Garment style: jacket
  • Size: x-large
  • Garment type: flame-resistant, welding, flame-retardant
  • Color: green
  • Material: cotton
  • Closure type: snaps
  • Hazardous protection level: non-hazardous protection
  • Chest size: 48 to 50 in.
  • Material weight: 9.00 oz
  • Number of pockets: 1
  • Country of origin: Mexico

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.