Learn More
CHEMSCIENCE ChemScience Steel H, Carbon, Manganese, Phosphorus, Sulfur, 5 ea
Catalog No. NC3917303
Encompass_Preferred
This steel H reference material is designed for the analysis of carbon, manganese, phosphorus, sulfur, silicon, nickel, and molybdenum concentrations. It serves as a certified standard for calibration and validation in industrial laboratory metallurgical testing applications.
  • Designed for elemental analysis of C, Mn, P, S, Si, Ni, and Mo
  • Suitable for calibration and quality control procedures in materials testing
  • Manufactured for consistent performance in laboratory environments
Catalog No. NC3917303 Supplier CHEMSCIENCE Supplier No. SI801700
This steel H reference material is designed for the analysis of carbon, manganese, phosphorus, sulfur, silicon, nickel, and molybdenum concentrations. It serves as a certified standard for calibration and validation in industrial laboratory metallurgical testing applications.
  • Designed for elemental analysis of C, Mn, P, S, Si, Ni, and Mo
  • Suitable for calibration and quality control procedures in materials testing
  • Manufactured for consistent performance in laboratory environments