Learn More
MSC Steel King Self-Closing Safety Gate, 48 in, Yellow, 1 ea
Catalog No. 5000781532
Encompass
This self-closing safety gate is designed to integrate with steel guard rail systems, providing enhanced versatility and safety in industrial environments. Constructed with heavy, 11-gauge corrugated steel rails, it is engineered to fit a 48-inch post-to-post centerline within a protective railing system.
  • Heavy, 11-gauge corrugated steel rails
  • Powder coat finish in safety yellow for high visibility, durability, and UV resistance
  • Versatile swing direction, easily changeable by re-positioning bolts
  • Gate opens to a 90-degree angle
  • Steel spring automatically closes gate after passage
  • Easy installation without cutting, welding, or drilling
  • Includes hinge assembly, two gate stops, necessary hardware, and end caps
  • Designed to fit a 48 in clear opening
  • Dimensions: 43-3/4 in wide x 39 in door height
Catalog No. 50-007-81532 Supplier MSC Supplier No. 41801234
This self-closing safety gate is designed to integrate with steel guard rail systems, providing enhanced versatility and safety in industrial environments. Constructed with heavy, 11-gauge corrugated steel rails, it is engineered to fit a 48-inch post-to-post centerline within a protective railing system.
  • Heavy, 11-gauge corrugated steel rails
  • Powder coat finish in safety yellow for high visibility, durability, and UV resistance
  • Versatile swing direction, easily changeable by re-positioning bolts
  • Gate opens to a 90-degree angle
  • Steel spring automatically closes gate after passage
  • Easy installation without cutting, welding, or drilling
  • Includes hinge assembly, two gate stops, necessary hardware, and end caps
  • Designed to fit a 48 in clear opening
  • Dimensions: 43-3/4 in wide x 39 in door height