Learn More
MSC Lakeside Medicine Cabinet, High Security, Wall Mounted, Steel, 1 ea
Catalog No. 502815680
Encompass
The Lakeside LHS-220 is a high security medicine and narcotics cabinet designed for wall mounting. Constructed from durable powder-coated steel, it provides secure storage for medications with an electronic keypad lock.
  • Comes standard with one fixed and one adjustable shelf
  • Electronic keypad lock with key override
  • Self-contained backup battery
  • Over 250 entry codes possible
  • Optional full audit trail capabilities
  • Made in the USA
Catalog No. 50-281-5680 Supplier MSC Supplier No. 28445773
The Lakeside LHS-220 is a high security medicine and narcotics cabinet designed for wall mounting. Constructed from durable powder-coated steel, it provides secure storage for medications with an electronic keypad lock.
  • Comes standard with one fixed and one adjustable shelf
  • Electronic keypad lock with key override
  • Self-contained backup battery
  • Over 250 entry codes possible
  • Optional full audit trail capabilities
  • Made in the USA