Learn More
Thermo Scientific Steel Needle, Pack of 5
Supplier:  Thermo Scientific 009500100437
Encompass
5 hollow sharp sterile steel disposable laboratory needles for piercing and transferring liquids
Catalog No. NC0827764