Learn More
Leica Biosystems Knife, 16 cm, Profile C, Steel - Reusable Microtome/Cryostat Knife
Catalog No. NC1238076
Encompass
This reusable steel knife features a profile C geometry and 16 cm length for precision sectioning of biological specimens. It is intended for routine histology and cryotomy applications, suitable for both paraffin-embedded and frozen tissue sections.
  • Precision ground steel blade with profile C geometry.
  • 16 cm length suitable for routine histology sectioning.
  • Designed for paraffin-embedded and frozen tissue sections.
  • Reusable design; eligible for resharpening services per manufacturer instructions.
  • Compatible with standard microtomes and cryostats.
Catalog No. NC1238076 Supplier Leica Biosystems Supplier No. 14021607100
This reusable steel knife features a profile C geometry and 16 cm length for precision sectioning of biological specimens. It is intended for routine histology and cryotomy applications, suitable for both paraffin-embedded and frozen tissue sections.
  • Precision ground steel blade with profile C geometry.
  • 16 cm length suitable for routine histology sectioning.
  • Designed for paraffin-embedded and frozen tissue sections.
  • Reusable design; eligible for resharpening services per manufacturer instructions.
  • Compatible with standard microtomes and cryostats.