Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra Biosciences Corp Steel rack for 25 mm tubes 3x8
SDP

Catalog No. 5000882063
Encompass

Steel rack for 25 mm tubes 3x8 90 mm

Catalog No. 50-008-82063 Supplier Integra Biosciences Corp Supplier No. 103078
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Steel rack for 25 mm tubes 3x8 90 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.