Learn More
Electron Microscopy Sciences Cut-Off Wheel Soft Steel Rc30-45
SDP
Catalog No. 50997139
Encompass_Preferred
Abrasive Cut-Off Wheels are offered for long life and are affordably priced for various sectioning requirements. Wheels are 9" (228.6 mm) diameter 0.062" (1.6 mm) thick; 1 1/4" (31 mm) arbor. Aluminum oxide abrasive cut-off wheel. Wheels are 9" (228.6mm) diameter 0.062" (1.6mm) thick; 1 1/4" (31mm) arbor. Soft Steel Rc 30-45.
Catalog No. 50-997-139 Supplier Electron Microscopy Sciences Supplier No. 5020915
Abrasive Cut-Off Wheels are offered for long life and are affordably priced for various sectioning requirements. Wheels are 9" (228.6 mm) diameter 0.062" (1.6 mm) thick; 1 1/4" (31 mm) arbor. Aluminum oxide abrasive cut-off wheel. Wheels are 9" (228.6mm) diameter 0.062" (1.6mm) thick; 1 1/4" (31mm) arbor. Soft Steel Rc 30-45.