Learn More
MSC PEC Tools Adjustable Hook Ruler, Carbon Steel, 6 in., English/Metric, 1 ea.
Catalog No. 5000516970
Encompass
These adjustable hook rulers are engineered for precise measurements from the end of a workpiece or within tight recesses where standard rules cannot reach. Designed for professional use, these tools eliminate the need for lining up the zero manually. Each rule is constructed from durable, fully hardened and tempered carbon steel with a non-glare, hard chrome finish for optimal readability.
  • Features an adjustable, reversible, and removable hook for versatile zero-point measurement
  • Precision-etched, black-filled graduations ensure long-lasting clarity
  • Industrial thickness of 0.040 in. provides a balance of rigidity and slim profile
  • Includes English and metric graduation options: 0.5 mm, 32nds, mm, and 64ths
Catalog No. 50-005-16970 Supplier MSC Supplier No. 01470418
These adjustable hook rulers are engineered for precise measurements from the end of a workpiece or within tight recesses where standard rules cannot reach. Designed for professional use, these tools eliminate the need for lining up the zero manually. Each rule is constructed from durable, fully hardened and tempered carbon steel with a non-glare, hard chrome finish for optimal readability.
  • Features an adjustable, reversible, and removable hook for versatile zero-point measurement
  • Precision-etched, black-filled graduations ensure long-lasting clarity
  • Industrial thickness of 0.040 in. provides a balance of rigidity and slim profile
  • Includes English and metric graduation options: 0.5 mm, 32nds, mm, and 64ths