Learn More
MSC PEC Tools Adjustable Hook Rule, Tool Steel, 450 mm, 0.5 mm Graduations, 1 ea.
Catalog No. 5000513577
Encompass
The PEC Tools 450mm Adjustable Hook Rule is designed for precise measurement applications, featuring a durable hardened and tempered tool steel construction. This professional-grade rule includes a hard chrome finish to provide a no-glare surface for improved readability in various lighting conditions.
  • Constructed from hardened and tempered tool steel for long-lasting durability
  • Features a non-glare hard chrome finish for clear visibility
  • Equipped with an adjustable hook for versatile measurement tasks
  • Metric graduations of 0.5 mm provided on both sides
  • Measures 0.040 in. in thickness
Catalog No. 50-005-13577 Supplier MSC Supplier No. 01470475
The PEC Tools 450mm Adjustable Hook Rule is designed for precise measurement applications, featuring a durable hardened and tempered tool steel construction. This professional-grade rule includes a hard chrome finish to provide a no-glare surface for improved readability in various lighting conditions.
  • Constructed from hardened and tempered tool steel for long-lasting durability
  • Features a non-glare hard chrome finish for clear visibility
  • Equipped with an adjustable hook for versatile measurement tasks
  • Metric graduations of 0.5 mm provided on both sides
  • Measures 0.040 in. in thickness