Learn More
Med Vet International Steel Stainless K-Wire, J0250P, 0.062in x 7in, 6 Count
Catalog No. 5000581584
Encompass_Preferred
K-Wire or Fine Orthopedic Wire. Highest quality stainless steel type 316LVM. U.S.A. made. Double Trocar Point.
Catalog No. 50-005-81584 Supplier Med Vet International Supplier No. JOR250P
K-Wire or Fine Orthopedic Wire. Highest quality stainless steel type 316LVM. U.S.A. made. Double Trocar Point.