Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502764616
Encompass_Preferred
The 3/4" x 0.020" x 1,200' Steel Strapping from Signode is a general purpose steel strapping made of high tensile steel. It has a break strength of 1,800 lbs, meets military spec MIL-STD-622F, and is chromated for moderate corrosion resistance. This steel strapping comes in coils, can be used for baling, bundling, reinforcing, and unitizing applications, and works with most Signode tensioning tools and seals. It provides a cost-effective solution for general purpose steel strapping needs.
Catalog No. 50-276-4616 Supplier Grainger Supplier No. 16P038
The 3/4" x 0.020" x 1,200' Steel Strapping from Signode is a general purpose steel strapping made of high tensile steel. It has a break strength of 1,800 lbs, meets military spec MIL-STD-622F, and is chromated for moderate corrosion resistance. This steel strapping comes in coils, can be used for baling, bundling, reinforcing, and unitizing applications, and works with most Signode tensioning tools and seals. It provides a cost-effective solution for general purpose steel strapping needs.