Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502764617
Encompass_Preferred
The Grainger standard steel strapping is available in widths from 5/8" to 2" and thicknesses from .020" to .031". It comes in coils from 200' to 1,200' long. The steel has a minimum tensile strength of 28,000 psi. The strapping is used for banding and bundling items like lumber, paper, and bricks. It works with standard seals and tools. This general purpose strapping provides economical, high-strength steel banding.
Catalog No. 50-276-4617 Supplier Grainger Supplier No. 16P041
The Grainger standard steel strapping is available in widths from 5/8" to 2" and thicknesses from .020" to .031". It comes in coils from 200' to 1,200' long. The steel has a minimum tensile strength of 28,000 psi. The strapping is used for banding and bundling items like lumber, paper, and bricks. It works with standard seals and tools. This general purpose strapping provides economical, high-strength steel banding.