Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502764618
Encompass_Preferred
The 0.023 in Steel Strapping from Grainger is a general purpose steel strapping made of low carbon steel with a light zinc coating. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is available in coil lengths ranging from 300 ft to 1,200 ft. The strapping has a standard width of 1/2 in and thicknesses of 0.023 in, 0.031 in, and 0.044 in. It can be used for bundling, palletizing, and reinforcing loads. The steel offers flexibility and high strength at low cost.
Catalog No. 50-276-4618 Supplier Grainger Supplier No. 16P043
The 0.023 in Steel Strapping from Grainger is a general purpose steel strapping made of low carbon steel with a light zinc coating. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is available in coil lengths ranging from 300 ft to 1,200 ft. The strapping has a standard width of 1/2 in and thicknesses of 0.023 in, 0.031 in, and 0.044 in. It can be used for bundling, palletizing, and reinforcing loads. The steel offers flexibility and high strength at low cost.