Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502764588
Encompass_Preferred
The 0.023 in Steel Strapping from Grainger is a general purpose steel strapping made of high tensile steel. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is available in coils. The strapping is 0.023 in thick, 1/2 in wide, and comes in coils of 100 ft, 500 ft, and 1000 ft lengths. It can be used for bundling, palletizing, and other general purpose applications. The product is sold by Grainger, product number 16P033.
Catalog No. 50-276-4588 Supplier Grainger Supplier No. 16P036
The 0.023 in Steel Strapping from Grainger is a general purpose steel strapping made of high tensile steel. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is available in coils. The strapping is 0.023 in thick, 1/2 in wide, and comes in coils of 100 ft, 500 ft, and 1000 ft lengths. It can be used for bundling, palletizing, and other general purpose applications. The product is sold by Grainger, product number 16P033.