Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502764589
Encompass_Preferred
The 0.029 in Steel Strapping from Grainger is a general purpose steel strapping made of high tensile steel. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is chromate coated for moderate corrosion resistance. This strapping comes in coils in multiple widths (1/2 in, 5/8 in, 3/4 in, 1 in) and lengths (300 ft, 600 ft). It can be used for general bundling, palletizing, and baling applications. The product is sold by Grainger, product number 16P043.
Catalog No. 50-276-4589 Supplier Grainger Supplier No. 16P040
The 0.029 in Steel Strapping from Grainger is a general purpose steel strapping made of high tensile steel. It has a break strength of 550 lbs, meets military spec MIL-STD-621D Type I, and is chromate coated for moderate corrosion resistance. This strapping comes in coils in multiple widths (1/2 in, 5/8 in, 3/4 in, 1 in) and lengths (300 ft, 600 ft). It can be used for general bundling, palletizing, and baling applications. The product is sold by Grainger, product number 16P043.