Learn More
Grainger Steel Strapping Portable Standard Duty
Catalog No. 502763743
Encompass_Preferred
The Grainger standard steel strapping is available in widths from 1/2" to 1-1/4" and thicknesses from .020" to .031". It comes in coils from 5,000' to 20,000' long. The finish is regular, wax, or epoxy coated. The strapping meets ASTM D3953 specifications. It is general use strapping suitable for unitizing, bundling, and palletizing applications. The working load limit ranges from 550 lbs to 2,200 lbs based on the dimensions. This carbon steel strapping provides good tensioning properties.
Catalog No. 50-276-3743 Supplier Grainger Supplier No. 16P039
The Grainger standard steel strapping is available in widths from 1/2" to 1-1/4" and thicknesses from .020" to .031". It comes in coils from 5,000' to 20,000' long. The finish is regular, wax, or epoxy coated. The strapping meets ASTM D3953 specifications. It is general use strapping suitable for unitizing, bundling, and palletizing applications. The working load limit ranges from 550 lbs to 2,200 lbs based on the dimensions. This carbon steel strapping provides good tensioning properties.