Learn More
Grainger Steel Strapping Standard Duty 3/4
Catalog No. 502764615
Encompass_Preferred
The 3/4" x 0.020" x 100' Steel Strapping from Grainger is a standard duty steel strapping made in the USA. It has a break strength of 1,250 lbs and meets military spec MIL-STD-621. The strapping is zinc coated for moderate corrosion resistance and has a smooth surface finish. It can be used for general bundling and baling applications. This product ships directly from Grainger.
Catalog No. 50-276-4615 Supplier Grainger Supplier No. 16P035
The 3/4" x 0.020" x 100' Steel Strapping from Grainger is a standard duty steel strapping made in the USA. It has a break strength of 1,250 lbs and meets military spec MIL-STD-621. The strapping is zinc coated for moderate corrosion resistance and has a smooth surface finish. It can be used for general bundling and baling applications. This product ships directly from Grainger.