Learn More
Grainger Steel Strapping Standard Duty 1/2
Catalog No. 502764543
Encompass_Preferred
The Grainger standard steel strapping is available in widths from 1/2" to 1-1/4" and thicknesses from .020" to .031". It comes in coils from 5,000' to 20,000' lengths. The finish is regular, waxed, or sealed. The strapping meets ASTM D3953 specifications. It is general use strapping suitable for bundling, palletizing, and other unitizing applications. The working load limit ranges from 500 lbs to 4,000 lbs based on the dimensions. This carbon steel strapping provides good tensioning with its high tensile strength.
Catalog No. 50-276-4543 Supplier Grainger Supplier No. 16P033
The Grainger standard steel strapping is available in widths from 1/2" to 1-1/4" and thicknesses from .020" to .031". It comes in coils from 5,000' to 20,000' lengths. The finish is regular, waxed, or sealed. The strapping meets ASTM D3953 specifications. It is general use strapping suitable for bundling, palletizing, and other unitizing applications. The working load limit ranges from 500 lbs to 4,000 lbs based on the dimensions. This carbon steel strapping provides good tensioning with its high tensile strength.