Learn More
Chemglass Life Sciences PTFE Coated Steel Support, Floor Model, 67in OAH, Holds 12L & 22L RBF
SDP
Catalog No. CG1539S100
Encompass_Preferred
Stainless Steel Support, Floor Model, 67in OAH, Holds 12L & 22L Round Bottom Flasks. Floor style stainless steel support stand made to accept 12 and 22 Liter CG-1539 half jacketed flasks. Stand is 67in overall height with a depth of 22in. Ring has a diameter of 8 1/2in I.D. and is 28in above floor level. Top of stand has a 39in reinforced riser to support overhead stirrers, clamps etc.
Catalog No. CG1539S100 Supplier Chemglass Life Sciences Supplier No. CG1539S100
Stainless Steel Support, Floor Model, 67in OAH, Holds 12L & 22L Round Bottom Flasks. Floor style stainless steel support stand made to accept 12 and 22 Liter CG-1539 half jacketed flasks. Stand is 67in overall height with a depth of 22in. Ring has a diameter of 8 1/2in I.D. and is 28in above floor level. Top of stand has a 39in reinforced riser to support overhead stirrers, clamps etc.