Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Steel Vernier Caliper
SCI_ED

Catalog No. S13602 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
120 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S13602 120 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Intended for ages 13 and up.
Catalog No. S13602 Supplier GSC Go Science Crazy Supplier No. 4VC34
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for ages 13 and up.

Measures with ease and precision in laboratory or classroom settings. The GSC International Steel Vernier Caliper is graduated with both metric and English units.

  • A useful measurement tool for laboratory and classroom settings
  • Roller adjustment and locking screw
  • Includes double scale graduations in both metric and English

Specifications

Calibration Type Roller adjustment; locking screw
For Use With (Application) Measurement in lab or classroom settings
Length (English) 5 in.
Length (Metric) 120 mm
Scale Type Metric and English
Material Steel
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.