Learn More
Grainger Vestil Work Platform, Steel, 36 in. x 48 in., 1,000 lb. Capacity
Catalog No. 502777331
Encompass_Preferred
This steel work platform provides a secure and stable elevated surface designed for use with a forklift. Constructed from durable steel, it features a non-slip tread plate floor and dual handrails on three sides to enhance worker safety during operation. The platform includes a patented automatic locking gate with an extra chain lock and a forklift tine lock assembly for secure attachment to forklift forks. This unit supports a maximum occupancy of two people and has a rated load capacity of 1,000 lb.
  • Maximum load capacity of 1,000 lb.
  • Patented automatic locking gate with extra chain lock for safety
  • Forklift tine lock assembly secures platform to forks
  • Durable steel construction with tread plate floor for non-slip grip
  • Dual handrails on three sides for worker protection
  • Compatible with a maximum occupancy of two people
  • Made in the USA
Catalog No. 50-277-7331 Supplier Grainger Supplier No. 173C74
This steel work platform provides a secure and stable elevated surface designed for use with a forklift. Constructed from durable steel, it features a non-slip tread plate floor and dual handrails on three sides to enhance worker safety during operation. The platform includes a patented automatic locking gate with an extra chain lock and a forklift tine lock assembly for secure attachment to forklift forks. This unit supports a maximum occupancy of two people and has a rated load capacity of 1,000 lb.
  • Maximum load capacity of 1,000 lb.
  • Patented automatic locking gate with extra chain lock for safety
  • Forklift tine lock assembly secures platform to forks
  • Durable steel construction with tread plate floor for non-slip grip
  • Dual handrails on three sides for worker protection
  • Compatible with a maximum occupancy of two people
  • Made in the USA