Learn More
Diagnostic Biosystems STEINER Steiner
Catalog No. 502407318
Encompass_Preferred
The Steiner Stain Kit (For Spirochetes) is designed for demonstrating Fungi, Helicobacter ylori, Legionella pneumophila, and Spirochete infected tissue. Kit may be used on formalin fixed, paraffin-embedded tissue.
Catalog No. 50-240-7318 Supplier Diagnostic Biosystems Supplier No. KT035
The Steiner Stain Kit (For Spirochetes) is designed for demonstrating Fungi, Helicobacter ylori, Legionella pneumophila, and Spirochete infected tissue. Kit may be used on formalin fixed, paraffin-embedded tissue.