Learn More
MSC Steiner ArcView Welding Curtain, 40 mil, 6x8 ft., 1 ea
Catalog No. 5000676361
Encompass
ArcView™ curtains provide protection from harmful UV rays and reduce visible flash when visibility is required. They are suitable for high abrasion applications.
  • Blocks most UV radiation
  • Forty mil thickness
  • Orange color
  • Meets California Fire Marshal requirements
  • Grommeted on all four sides, 12 in. apart
  • Flame, water, and mildew resistant
Catalog No. 50-006-76361 Supplier MSC Supplier No. 40742678
ArcView™ curtains provide protection from harmful UV rays and reduce visible flash when visibility is required. They are suitable for high abrasion applications.
  • Blocks most UV radiation
  • Forty mil thickness
  • Orange color
  • Meets California Fire Marshal requirements
  • Grommeted on all four sides, 12 in. apart
  • Flame, water, and mildew resistant