Learn More
MSC Steiner Flame Resistant Jacket, Medium, Cotton, Snaps Closure, 1 ea
Catalog No. 5000768147
Encompass
This flame resistant/retardant jacket is constructed from cotton and features a snap closure, providing essential protection in hazardous work environments.
  • Medium size
  • Green color
  • Cotton material
  • Snaps closure
  • Chest size: 44-46 in.
  • Material weight: 12.0 oz
  • Rot resistant
  • Anti-static
  • Mildew resistant
  • Leather reinforced
  • Non-reflective snaps
  • One pocket
  • Country of origin: Philippines
Catalog No. 50-007-68147 Supplier MSC Supplier No. 04416491
This flame resistant/retardant jacket is constructed from cotton and features a snap closure, providing essential protection in hazardous work environments.
  • Medium size
  • Green color
  • Cotton material
  • Snaps closure
  • Chest size: 44-46 in.
  • Material weight: 12.0 oz
  • Rot resistant
  • Anti-static
  • Mildew resistant
  • Leather reinforced
  • Non-reflective snaps
  • One pocket
  • Country of origin: Philippines