Learn More
MSC Steiner FR Cotton Jacket, 9 oz, 30 in, 1 ea
Catalog No. 5000767100
Encompass
This Steiner FR cotton jacket offers cool, comfortable, and washable protection for light welding and from light sparks, meeting ASTM D6413-10 standards.
  • Relaxed fit design for all-day wearability
  • Adjustable snap wrist and large inside pocket
  • Leather reinforced, black anodized non-reflective snaps
  • Weldlite™ high quality fabric is anti-static, rot and mildew resistant
  • Maintains flame retardancy for up to 50 home or 25 commercial washings
  • FR patch and soapstone pocket on left sleeve
Catalog No. 50-007-67100 Supplier MSC Supplier No. 55579759
This Steiner FR cotton jacket offers cool, comfortable, and washable protection for light welding and from light sparks, meeting ASTM D6413-10 standards.
  • Relaxed fit design for all-day wearability
  • Adjustable snap wrist and large inside pocket
  • Leather reinforced, black anodized non-reflective snaps
  • Weldlite™ high quality fabric is anti-static, rot and mildew resistant
  • Maintains flame retardancy for up to 50 home or 25 commercial washings
  • FR patch and soapstone pocket on left sleeve