Learn More
MSC Steiner FR Hybrid Jacket, Cotton/Leather, 9 oz, Small, 1 ea
Catalog No. 5000787115
Encompass
The Steiner 1230 Weldlite Plus Hybrid Jacket is designed to offer a combination of comfort and protection for demanding applications. Its torso is constructed from 9 oz flame retardant 100% cotton Weldlite™ fabric, complemented by rugged side split cowhide gray leather sleeves.
  • Torso made from 9 oz flame retardant 100% cotton Weldlite™ fabric
  • Rugged side split cowhide gray leather sleeves
  • Relaxed fit design
  • Adjustable snap wrist
  • Large inside pocket
  • Leather reinforced, black anodized non-reflective snaps
  • Weldlite™ high quality fabric is anti-static, rot, and mildew resistant
  • Sleeves are sewn with Kevlar® thread
  • Bar tacked at stress points
  • Soapstone pocket on each sleeve
Catalog No. 50-007-87115 Supplier MSC Supplier No. 32133498
The Steiner 1230 Weldlite Plus Hybrid Jacket is designed to offer a combination of comfort and protection for demanding applications. Its torso is constructed from 9 oz flame retardant 100% cotton Weldlite™ fabric, complemented by rugged side split cowhide gray leather sleeves.
  • Torso made from 9 oz flame retardant 100% cotton Weldlite™ fabric
  • Rugged side split cowhide gray leather sleeves
  • Relaxed fit design
  • Adjustable snap wrist
  • Large inside pocket
  • Leather reinforced, black anodized non-reflective snaps
  • Weldlite™ high quality fabric is anti-static, rot, and mildew resistant
  • Sleeves are sewn with Kevlar® thread
  • Bar tacked at stress points
  • Soapstone pocket on each sleeve