Learn More
MSC Steiner FR Jacket, 2X-Large, Cotton and Leather, Snaps, 1 ea
Catalog No. 5000673812
Encompass
This flame resistant/retardant jacket offers robust protection for welding applications, crafted from a durable blend of cotton and leather with a secure snaps closure.
  • Garment style: Jacket
  • Size: 2X-Large
  • Flame resistant/retardant garment type
  • Color: Blue
  • Material: Cotton and leather
  • Closure type: Snaps
  • Hazardous protection level: ASTM D6413-10
  • Material weight: 9.0 oz
  • Features bar tacked at stress points
  • Leather reinforced construction
  • Non-reflective snaps
  • One pocket for storage
  • Country of origin: Philippines
Catalog No. 50-006-73812 Supplier MSC Supplier No. 55580237
This flame resistant/retardant jacket offers robust protection for welding applications, crafted from a durable blend of cotton and leather with a secure snaps closure.
  • Garment style: Jacket
  • Size: 2X-Large
  • Flame resistant/retardant garment type
  • Color: Blue
  • Material: Cotton and leather
  • Closure type: Snaps
  • Hazardous protection level: ASTM D6413-10
  • Material weight: 9.0 oz
  • Features bar tacked at stress points
  • Leather reinforced construction
  • Non-reflective snaps
  • One pocket for storage
  • Country of origin: Philippines