Learn More
MSC Steiner Leather Welding Blanket, 4x3 ft, 0.06-0.08 in, 1 ea
Catalog No. 5000781909
Encompass
This leather welding blanket is designed for protection in welding applications. It measures 4 ft. high by 3 ft. wide and has a thickness of 0.06 to 0.08 in.
  • Made from leather material
  • Provides protection for welding applications
  • Measures 4 ft. high and 3 ft. wide
  • Thickness ranges from 0.06 to 0.08 in.
  • Material weight of 4.00 oz/sq. yd.
  • Color: Rust
  • Does not include grommets
  • Manufactured in China
Catalog No. 50-007-81909 Supplier MSC Supplier No. 55578157
This leather welding blanket is designed for protection in welding applications. It measures 4 ft. high by 3 ft. wide and has a thickness of 0.06 to 0.08 in.
  • Made from leather material
  • Provides protection for welding applications
  • Measures 4 ft. high and 3 ft. wide
  • Thickness ranges from 0.06 to 0.08 in.
  • Material weight of 4.00 oz/sq. yd.
  • Color: Rust
  • Does not include grommets
  • Manufactured in China