Learn More
MSC Steiner MIG Welding Gloves, Grain Goatskin/Split Cowhide, 2X-Large, 1 Pair
Catalog No. 5000767127
Encompass
These MIG welding gloves are engineered for extreme comfort, dexterity, and protection during MIG gun operation. They feature a snug-fit style to allow for the dexterity needed for trigger and gun handling, making them ideal for welding and grinding applications.
  • Premium heavyweight pearl grain goatskin palm for dexterity
  • Durable split cowhide back with reinforced thumb saddle
  • Fully cotton fleece lined for added comfort
  • Seamless index finger for enhanced feel and protection
  • Straight thumb for best gun grip
  • Elastic gathered back snugs hands for comfortable fit
  • Durable lock stitching with flame resistant thread
  • Gauntlet cuff provides extended wrist protection
Catalog No. 50-007-67127 Supplier MSC Supplier No. 39952239
These MIG welding gloves are engineered for extreme comfort, dexterity, and protection during MIG gun operation. They feature a snug-fit style to allow for the dexterity needed for trigger and gun handling, making them ideal for welding and grinding applications.
  • Premium heavyweight pearl grain goatskin palm for dexterity
  • Durable split cowhide back with reinforced thumb saddle
  • Fully cotton fleece lined for added comfort
  • Seamless index finger for enhanced feel and protection
  • Straight thumb for best gun grip
  • Elastic gathered back snugs hands for comfortable fit
  • Durable lock stitching with flame resistant thread
  • Gauntlet cuff provides extended wrist protection