Learn More
Newcomer Supply Steiner-Chapman Modified Silver Stain Kit, 250 ml kit - No Uranyl Nitrate!
SDP
Catalog No. NC2102291
Encompass
Newcomer Supply Steiner-Chapman Modified Silver Stain Kit procedure, with included microwave modification, is used for staining spirochetes, Helicobacter pylori, Legionella pneumophila, other nonfilamentous bacteria and fungus. This modified method eliminates the use of Uranyl Nitrate and its regulatory and disposal requirements.
This item is not returnable. View return policy
Catalog No. NC2102291 Supplier Newcomer Supply Supplier No. 9172A
Newcomer Supply Steiner-Chapman Modified Silver Stain Kit procedure, with included microwave modification, is used for staining spirochetes, Helicobacter pylori, Legionella pneumophila, other nonfilamentous bacteria and fungus. This modified method eliminates the use of Uranyl Nitrate and its regulatory and disposal requirements.