Learn More
MSC Steiner Technologies Thread Plug Gage, UNC Nr.1-64, 2B, GO/NO GO
Catalog No. 5000529398
Encompass
A GO/NO GO Thread Plug Gage from Steiner Technologies designed for verifying internal threads with UNC Nr.1-64 dimensions and 2B tolerance. This precision measuring instrument ensures adherence to thread specifications for quality control and manufacturing processes.
  • Designed for internal thread verification.
  • Ensures precise adherence to UNC Nr.1-64 thread specifications.
  • Features 2B tolerance for accurate gaging.
  • Uncoated for general purpose applications.
  • Essential tool for quality control in manufacturing.
Catalog No. 50-005-29398 Supplier MSC Supplier No. 05361654
A GO/NO GO Thread Plug Gage from Steiner Technologies designed for verifying internal threads with UNC Nr.1-64 dimensions and 2B tolerance. This precision measuring instrument ensures adherence to thread specifications for quality control and manufacturing processes.
  • Designed for internal thread verification.
  • Ensures precise adherence to UNC Nr.1-64 thread specifications.
  • Features 2B tolerance for accurate gaging.
  • Uncoated for general purpose applications.
  • Essential tool for quality control in manufacturing.