Learn More
MSC Steiner Vinyl Welding Curtain, 10x8 ft, Amber, 1 ea
Catalog No. 5000802463
Encompass
This welding curtain kit features durable vinyl strips, providing a protective barrier for welding applications. It is designed to create a safe work environment by containing sparks and glare.
  • Constructed from durable vinyl material
  • Measures 10 ft. wide by 8 ft. high
  • Features a thickness of 0.08 in.
  • Supplied in an amber color
  • Includes pipe mount clamping hardware for installation
  • Material weight of 72.00 oz/sq. yd.
  • Manufactured in the United States
Catalog No. 50-008-02463 Supplier MSC Supplier No. 02037166
This welding curtain kit features durable vinyl strips, providing a protective barrier for welding applications. It is designed to create a safe work environment by containing sparks and glare.
  • Constructed from durable vinyl material
  • Measures 10 ft. wide by 8 ft. high
  • Features a thickness of 0.08 in.
  • Supplied in an amber color
  • Includes pipe mount clamping hardware for installation
  • Material weight of 72.00 oz/sq. yd.
  • Manufactured in the United States