Learn More
MSC Steiner Welding Curtain, 14 mil, Orange, 6x6 ft, 1 ea
Catalog No. 5000792994
Encompass
This ArcView™ welding curtain is made from tinted transparent vinyl, designed to offer protection from harmful UV rays and reduce visible flash during welding operations. It is also resistant to flame, water, and mildew, ensuring durability and safety in various environments.
  • Blocks most UV radiation
  • Provides protection from harmful UV rays and reduces visible flash
  • Has 14 mil thickness
  • Orange color
  • Meets California Fire Marshal requirements
  • Features 3/8 in. brass grommets on all four sides, spaced 12 in. apart
  • Resistant to flame, water, and mildew
Catalog No. 50-007-92994 Supplier MSC Supplier No. 84339696
This ArcView™ welding curtain is made from tinted transparent vinyl, designed to offer protection from harmful UV rays and reduce visible flash during welding operations. It is also resistant to flame, water, and mildew, ensuring durability and safety in various environments.
  • Blocks most UV radiation
  • Provides protection from harmful UV rays and reduces visible flash
  • Has 14 mil thickness
  • Orange color
  • Meets California Fire Marshal requirements
  • Features 3/8 in. brass grommets on all four sides, spaced 12 in. apart
  • Resistant to flame, water, and mildew