Learn More
MSC Steiner Welding Curtain, 6x6 ft, Window, 1 ea
Catalog No. 5000782554
Encompass
This welding curtain is designed to offer robust protection during welding operations. Constructed from silicone-coated fiberglass, it features a unique Weld-View Window for enhanced visibility, alongside high heat and abrasion resistance.
  • Made from silicone-coated fiberglass
  • Features a weld-view window with hook and loop attachment for easy replacement
  • Withstands temperatures up to 1,000 degrees F
  • Measures 6 ft wide by 6 ft high
  • Material weight of 16 oz/sq yd and 0.41 mm thickness
  • Equipped with grommets for secure fastening
  • Abrasion and corrosion resistant
  • California Fire Marshal approved
Catalog No. 50-007-82554 Supplier MSC Supplier No. 70228192
This welding curtain is designed to offer robust protection during welding operations. Constructed from silicone-coated fiberglass, it features a unique Weld-View Window for enhanced visibility, alongside high heat and abrasion resistance.
  • Made from silicone-coated fiberglass
  • Features a weld-view window with hook and loop attachment for easy replacement
  • Withstands temperatures up to 1,000 degrees F
  • Measures 6 ft wide by 6 ft high
  • Material weight of 16 oz/sq yd and 0.41 mm thickness
  • Equipped with grommets for secure fastening
  • Abrasion and corrosion resistant
  • California Fire Marshal approved