Learn More
MSC Steiner, Welding Curtain, ArcView, 40 mil, 4x6 ft, 1 ea
Catalog No. 5000795171
Encompass
ArcView™ curtains provide protection from harmful UV rays and reduce the visible flash when visibility is required. They are suitable for high abrasion applications.
  • Blocks most UV radiation
  • Features 40 mil thickness
  • Has orange color
  • Meets California Fire Marshal requirements
  • Grommeted on all four sides, 12 in apart
  • Flame, water, and mildew resistant
Catalog No. 50-007-95171 Supplier MSC Supplier No. 40742694
ArcView™ curtains provide protection from harmful UV rays and reduce the visible flash when visibility is required. They are suitable for high abrasion applications.
  • Blocks most UV radiation
  • Features 40 mil thickness
  • Has orange color
  • Meets California Fire Marshal requirements
  • Grommeted on all four sides, 12 in apart
  • Flame, water, and mildew resistant