Learn More
MSC Steiner Welding Curtain Roll, 75 x 5 ft, 1 ea
Catalog No. 5000782785
Encompass
This opaque welding curtain roll provides protection against light sparks and spatter on a vertical plane. Made from flame, mildew, and water-resistant cotton duck, it is suitable for general-purpose welding applications.
  • Opaque design blocks flash and harmful rays
  • Flame, mildew, and water-resistant properties
  • Withstands temperatures up to 150 degrees F
  • Equipped with brass grommets on 12-inch centers along one side
  • California Fire Marshal Approved for safety
Catalog No. 50-007-82785 Supplier MSC Supplier No. 93215838
This opaque welding curtain roll provides protection against light sparks and spatter on a vertical plane. Made from flame, mildew, and water-resistant cotton duck, it is suitable for general-purpose welding applications.
  • Opaque design blocks flash and harmful rays
  • Flame, mildew, and water-resistant properties
  • Withstands temperatures up to 150 degrees F
  • Equipped with brass grommets on 12-inch centers along one side
  • California Fire Marshal Approved for safety