Learn More
MSC Steiner Welding Curtain, Vinyl Polyester, 6x8ft., 1 ea
Catalog No. 5000673952
Encompass
This vinyl laminated polyester welding curtain provides a protective barrier for welding environments. Designed for durability, it features grommets for easy installation and is suitable for various industrial applications.
  • Made from durable vinyl laminated polyester
  • Features grommets for secure hanging
  • Measures 6 ft. wide by 8 ft. high
  • Material weight of 13 oz/sq. yd.
  • Black finish
Catalog No. 50-006-73952 Supplier MSC Supplier No. 50691500
This vinyl laminated polyester welding curtain provides a protective barrier for welding environments. Designed for durability, it features grommets for easy installation and is suitable for various industrial applications.
  • Made from durable vinyl laminated polyester
  • Features grommets for secure hanging
  • Measures 6 ft. wide by 8 ft. high
  • Material weight of 13 oz/sq. yd.
  • Black finish