Learn More
MSC Steiner Welding Curtain, Weld-View, 6 x 8 ft, 1 ea
Catalog No. 5000794986
Encompass
This welding curtain is constructed from durable RedGlass™ coated fiberglass, providing robust protection for welding environments. It effectively shields against sparks, intense heat, and harmful UV radiation, while its integrated Weld-View window enables safe observation of welding operations.
  • Opaque curtain with replaceable tinted transparent viewing window
  • Superior heat, spark, and abrasion resistance
  • Blocks flash and harmful UV rays
  • Heavy duty frame with grommets on all four sides
  • Withstands temperatures up to 1,000 degrees F
  • California Fire Marshal approved
Catalog No. 50-007-94986 Supplier MSC Supplier No. 70228176
This welding curtain is constructed from durable RedGlass™ coated fiberglass, providing robust protection for welding environments. It effectively shields against sparks, intense heat, and harmful UV radiation, while its integrated Weld-View window enables safe observation of welding operations.
  • Opaque curtain with replaceable tinted transparent viewing window
  • Superior heat, spark, and abrasion resistance
  • Blocks flash and harmful UV rays
  • Heavy duty frame with grommets on all four sides
  • Withstands temperatures up to 1,000 degrees F
  • California Fire Marshal approved